siRNA Details
Gene Symbol: 2310002L09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027104.3 | NP_081380.2 | 2 | 521 | ||
NM_027981.1 | NP_082257.1 | 2 | 521 | |||
GenBank mRNA | AK009097.1 | - | 2 | 522 | ||
AK009124.1 | - | 2 | 521 | |||
BC115483.1 | - | 2 | 465 | 40100595 | ||
BC115484.1 | - | 2 | 465 | 40100599 |
