siRNA Details
Gene Symbol: Apol9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168660.1 | NP_001162131.1 | 3 | 701 | ||
NM_173743.4 | NP_776104.1 | 3 | 652 | |||
GenBank mRNA | AK009386.1 | - | 3 | 702 | ||
AK050167.1 | - | 3 | 654 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
