siRNA Details
Gene Symbol: Tmem106b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027992.3 | NP_082268.2 | 3 | 417 | ||
GenBank mRNA | AK009646.1 | - | 3 | 320 | ||
AK018015.1 | - | 3 | 324 | |||
AK148477.1 | - | 3 | 339 | |||
BC050246.1 | - | 3 | 334 | 6313498 |
