siRNA Details
Gene Symbol: Cand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027994.1 | NP_082270.1 | 10 | 2800 | ||
GenBank mRNA | AK129225.1 | - | 10 | 2761 | ||
BC057457.1 | - | 3 | 1443 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
