siRNA Details
Gene Symbol: Ppapdc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293703.1 | NP_001280632.1 | 4 | 338 | ||
NM_028000.1 | NP_082276.1 | 4 | 338 | |||
GenBank mRNA | AA184457.1 | - | 4 | 338 | ||
AK007542.1 | - | 2 | 299 | |||
AK009455.1 | - | 4 | 305 | |||
AK144593.1 | - | 4 | 323 | |||
BB857717.1 | - | 4 | 339 | |||
BC023111.1 | - | 4 | 285 | |||
BC039626.1 | - | 3 | 153 | |||
BC062173.1 | - | 4 | 271 | |||
BC089556.1 | - | 4 | 282 | |||
BC099489.1 | - | 4 | 289 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
