siRNA Details
Gene Symbol: Jsrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028001.3 | NP_082277.2 | 7 | 704 | ||
GenBank mRNA | AK009016.1 | - | 7 | 698 | ||
AK009578.1 | - | 7 | 919 | |||
BC104136.1 | - | 6 | 618 | |||
BC104137.1 | - | 6 | 619 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
