siRNA Details
Gene Symbol: Tmem123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133739.2 | NP_598500.1 | 3 | 383 | ||
GenBank mRNA | AK150361.1 | - | 3 | 331 | ||
AK150365.1 | - | 4 | 411 | |||
AK151044.1 | - | 3 | 351 | |||
AK152180.1 | - | 3 | 331 | |||
AK152423.1 | - | 3 | 296 | |||
AK153122.1 | - | 3 | 296 | |||
AK153378.1 | - | 4 | 411 | |||
AK153539.1 | - | 3 | 331 | |||
AK155767.1 | - | 3 | 351 | |||
AK157001.1 | - | 4 | 308 | |||
AK162610.1 | - | 3 | 379 | |||
AK165671.1 | - | 3 | 350 | |||
AK166444.1 | - | 3 | 296 | |||
AK170823.1 | - | 3 | 342 | |||
AK172352.1 | - | 3 | 312 | |||
AK172553.1 | - | 3 | 312 | |||
AK172701.1 | - | 3 | 312 | |||
AK211851.1 | - | 1 | 38 | |||
BC010292.1 | - | 3 | 332 | 3153174 | ||
BC086319.1 | - | 3 | 340 | 5293409 | ||
BY259389.1 | - | 3 | 384 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
