siRNA Details
Gene Symbol: Cers5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028015.2 | NP_082291.1 | 3 | 489 | ||
GenBank mRNA | AK010241.1 | - | 3 | 417 | ||
AK075694.1 | - | 3 | 409 | |||
AK145589.1 | - | 3 | 397 | |||
AK145762.1 | - | 3 | 411 | |||
AK146220.1 | - | 3 | 411 | |||
AK146417.1 | - | 3 | 380 | |||
AK219835.1 | - | 3 | 199 | |||
AY029533.1 | - | 3 | 409 | |||
BC043059.1 | - | 436 | 6419704 | |||
BC046797.1 | - | 3 | 489 | 6403129 |
