siRNA Details
Gene Symbol: Nanog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289828.1 | NP_001276757.1 | 4 | 1993 | ||
NM_001289830.1 | NP_001276759.1 | 5 | 1793 | |||
NM_001289831.1 | NP_001276760.1 | 5 | 1777 | |||
NM_028016.3 | NP_082292.1 | 4 | 1996 | |||
GenBank mRNA | AB093574.1 | - | 4 | 1967 | ||
AF507043.1 | - | 4 | 1968 | |||
AK010332.1 | - | 4 | 1968 | |||
AK167259.1 | - | 4 | 1968 | |||
AY278951.1 | - | 4 | 1845 | |||
AY455282.1 | - | 5 | 1777 | |||
AY455285.1 | - | 5 | 1793 | |||
BC083105.1 | - | 1 | 876 | |||
BY382803.1 | - | 1 | 211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
