siRNA Details
Gene Symbol: Nanog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289828.1 | NP_001276757.1 | 4 | 2020 | ||
NM_001289830.1 | NP_001276759.1 | 5 | 1820 | |||
NM_001289831.1 | NP_001276760.1 | 5 | 1804 | |||
NM_028016.3 | NP_082292.1 | 4 | 2023 | |||
GenBank mRNA | AB093574.1 | - | 4 | 1994 | ||
AF507043.1 | - | 4 | 1995 | |||
AK010332.1 | - | 4 | 1995 | |||
AK167259.1 | - | 4 | 1995 | |||
AY278951.1 | - | 4 | 1872 | |||
AY455282.1 | - | 5 | 1804 | |||
AY455285.1 | - | 5 | 1820 | |||
BC083105.1 | - | 1 | 903 | |||
BY382803.1 | - | 1 | 238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
