siRNA Details
Gene Symbol: 2410016O06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023633.3 | NP_076122.2 | 1 | 1845 | ||
GenBank mRNA | AB041553.1 | - | 1 | 1784 | ||
AK010521.1 | - | 1 | 951 | |||
AK080870.1 | - | 1 | 1791 | |||
BC052152.2 | - | 1 | 1759 | |||
BC059809.1 | - | 1 | 1796 | |||
BC138094.1 | - | 1 | 1845 | 8861113 |
