siRNA Details
Gene Symbol: 2410016O06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023633.3 | NP_076122.2 | 1 | 1668 | ||
GenBank mRNA | AB041553.1 | - | 1 | 1607 | ||
AK010521.1 | - | 1 | 774 | |||
AK080870.1 | - | 1 | 1614 | |||
AK213032.1 | - | 1 | 255 | |||
BC052152.2 | - | 1 | 1582 | |||
BC059809.1 | - | 1 | 1619 | |||
BC138094.1 | - | 1 | 1668 | 8861113 |
