siRNA Details
Gene Symbol: Esco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028039.2 | NP_082315.3 | 3 | 841 | ||
GenBank mRNA | AK083728.1 | - | 1 | 620 | ||
AK144685.1 | - | 3 | 797 | |||
AK166457.1 | - | 3 | 789 | |||
BC033303.1 | - | 3 | 756 | 3675568 | ||
BC071275.1 | - | 3 | 774 | 6331551 | ||
CF730483.1 | - | 1 | 361 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
