siRNA Details
Gene Symbol: Rpusd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028040.2 | NP_082316.1 | 1 | 164 | ||
GenBank mRNA | AK010322.1 | - | 1 | 181 | ||
AK156574.1 | - | 1 | 181 | |||
BC061256.1 | - | 1 | 165 | 30251586 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
