siRNA Details
Gene Symbol: Cnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028044.2 | NP_082320.1 | 7 | 958 | ||
GenBank mRNA | AK005460.1 | - | 7 | 960 | ||
AK051121.1 | - | 6 | 1176 | |||
AK051628.1 | - | 7 | 976 | |||
AK081672.1 | - | 7 | 873 | |||
AK159555.1 | - | 7 | 1003 | |||
AK159871.1 | - | 7 | 988 | |||
AK166578.1 | - | 7 | 966 | |||
AK167201.1 | - | 7 | 979 | |||
AK167613.1 | - | 7 | 976 | |||
AK169021.1 | - | 7 | 983 | |||
AK169098.1 | - | 7 | 1003 | |||
AK169410.1 | - | 7 | 933 | |||
AK205558.1 | - | 1 | 63 | |||
BC005788.1 | - | 3 | 285 | |||
BC039643.1 | - | 3 | 353 | |||
BC055711.1 | - | 7 | 959 | 6415164 | ||
BC085268.1 | - | 7 | 736 | 6487559 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
