siRNA Details
Gene Symbol: Slc35f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028060.3 | NP_082336.3 | 5 | 968 | ||
GenBank mRNA | AK005182.1 | - | 5 | 764 | ||
BC019202.1 | - | 5 | 769 | |||
BC055843.1 | - | 5 | 965 | 3991126 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
