siRNA Details
Gene Symbol: Mccc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023644.4 | NP_076133.3 | 15 | 1825 | ||
GenBank mRNA | AF310338.1 | - | 15 | 1746 | ||
AK007782.1 | - | 15 | 1778 | |||
AK031072.1 | - | 15 | 1786 | |||
AK032203.1 | - | 15 | 1768 | |||
AK141040.1 | - | 1 | 675 | |||
AK168589.1 | - | 15 | 1714 | |||
BC021382.1 | - | 15 | 1789 | 5042993 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
