siRNA Details
Gene Symbol: Mccc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023644.4 | NP_076133.3 | 13, 14 | 1711 | ||
GenBank mRNA | AF310338.1 | - | 13, 14 | 1632 | ||
AK007782.1 | - | 13, 14 | 1664 | |||
AK031072.1 | - | 13, 14 | 1672 | |||
AK032203.1 | - | 13, 14 | 1654 | |||
AK168589.1 | - | 13, 14 | 1600 | |||
AK201441.1 | - | 1, 2 | 155 | |||
BC021382.1 | - | 13, 14 | 1675 | 5042993 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
