siRNA Details
Gene Symbol: Kdelc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023645.3 | NP_076134.1 | 8 | 1597 | ||
GenBank mRNA | AJ404004.1 | - | 8 | 1359 | ||
AK007835.1 | - | 8 | 1590 | |||
AK035899.1 | - | 8 | 1598 | |||
AK053315.1 | - | 7 | 1380 | |||
AK079701.1 | - | 8 | 1590 | |||
AK170300.1 | - | 8 | 1626 | |||
BC023141.1 | - | 7 | 1697 | 4018698 |
