siRNA Details
Gene Symbol: Tmub2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008158.1 | - | 3 | 688 | ||
AK132118.1 | - | 3 | 733 | |||
BC016187.1 | - | 2 | 674 | |||
BC029841.1 | - | 2 | 991 | 4209499 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
