siRNA Details
Gene Symbol: Sapcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081085.2 | NP_001074554.1 | 6 | 1534 | ||
NM_001290407.1 | NP_001277336.1 | 5 | 1432 | |||
GenBank mRNA | AB197927.1 | - | 5 | 1432 | ||
AK008577.1 | - | 5 | 1129 | |||
AK154143.1 | - | 6 | 1245 | |||
AK163973.1 | - | 6 | 753 | |||
AK164615.1 | - | 6 | 1209 | |||
AK166625.1 | - | 6 | 1209 | |||
BC070477.1 | - | 4 | 1221 | 30656854 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
