siRNA Details
Gene Symbol: Pigx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111025.1 | NP_001104495.1 | 6 | 810 | ||
NM_024464.3 | NP_077784.2 | 6 | 816 | |||
GenBank mRNA | AK008583.2 | - | 6 | 818 | ||
BC002202.1 | - | 4 | 460 | 3489499 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
