siRNA Details
Gene Symbol: Pigx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111025.1 | NP_001104495.1 | 4 | 505 | ||
NM_024464.3 | NP_077784.2 | 4 | 511 | |||
GenBank mRNA | AK008583.2 | - | 4 | 513 | ||
BC002202.1 | - | 2 | 155 | 3489499 | ||
BX525135.1 | - | 4 | 478 | |||
BY708475.1 | - | 4 | 512 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
