siRNA Details
Gene Symbol: Osgepl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285839.1 | NP_001272768.1 | 5 | 1272 | ||
GenBank mRNA | AK011265.1 | - | 5 | 1220 | ||
AK045669.1 | - | 5 | 1234 | |||
AK137332.1 | - | 5 | 1223 | |||
BC030671.1 | - | 5 | 1237 | |||
BC038910.1 | - | 3 | 598 | |||
BC051211.1 | - | 2 | 648 | |||
BC058172.1 | - | 5 | 1235 | 5012054 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
