siRNA Details
Gene Symbol: Jmjd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028101.4 | NP_082377.2 | 2, 3 | 214 | ||
GenBank mRNA | AK011308.1 | - | 2, 3 | 211 | ||
AK148079.1 | - | 2, 3 | 192 | |||
AK155810.1 | - | 2, 3 | 216 | |||
AK172073.1 | - | 2, 3 | 211 | |||
BC010800.1 | - | 2, 3 | 176 | 4235072 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
