siRNA Details
Gene Symbol: Ddhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028102.1 | NP_082378.1 | 2 | 233 | ||
GenBank mRNA | AK008529.1 | - | 2 | 219 | ||
AK035532.1 | - | 2 | 285 | |||
AK039979.1 | - | 1 | 1250 | |||
AK079311.1 | - | 2 | 281 | |||
AK173010.1 | - | 2 | 185 | |||
BC046229.1 | - | 1 | 153 | |||
BC118962.2 | - | 2 | 468 | 40095026 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
