siRNA Details
Gene Symbol: Ppp1r14d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290796.1 | NP_001277725.1 | 3 | 441 | ||
NM_028104.4 | NP_082380.1 | 2 | 376 | |||
GenBank mRNA | AF408400.2 | - | 2 | 375 | ||
AK008348.1 | - | 2 | 334 | |||
AY179331.1 | - | 3 | 428 | |||
BC024547.1 | - | 2 | 375 | 4989982 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
