siRNA Details
Gene Symbol: Naa50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028108.3 | NP_082384.1 | 3, 4 | 554 | ||
GenBank mRNA | AK011160.1 | - | 3, 4 | 480 | ||
AK135658.1 | - | 3, 4 | 497 | |||
AK145345.1 | - | 3, 4 | 482 | |||
AK156791.1 | - | 3, 4 | 500 | |||
AK167141.1 | - | 4, 5 | 674 | |||
AK168394.1 | - | 3, 4 | 525 | |||
AK184762.1 | - | 2, 3 | 250 | |||
AK195449.1 | - | 2, 3 | 250 | |||
AK198408.1 | - | 3, 4 | 323 | |||
AK219650.1 | - | 2, 3 | 250 | |||
BC004086.1 | - | 3, 4 | 466 | |||
BC023275.1 | - | 1, 2 | 13 | |||
BC055846.1 | - | 3, 4 | 319 | 3600545 | ||
BC057117.1 | - | 3, 4 | 513 | 6409354 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
