siRNA Details
Gene Symbol: Adpgk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028121.2 | NP_082397.1 | 7 | 1392 | ||
GenBank mRNA | AK011434.1 | - | 7 | 1357 | ||
AK149942.1 | - | 7 | 1361 | |||
AK162169.1 | - | 7 | 1346 | |||
AK167634.1 | - | 7 | 1390 | |||
AK169590.1 | - | 7 | 1356 | |||
BC021526.1 | - | 7 | 1376 | 5353637 | ||
BC094657.1 | - | 6 | 4064 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
