siRNA Details
Gene Symbol: Tdrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173744.4 | NP_776105.1 | 2 | 322 | ||
GenBank mRNA | AK077810.1 | - | 2 | 236 | ||
AK166875.1 | - | 2 | 233 | |||
AK166972.1 | - | 2 | 229 | |||
BC099613.1 | - | 2 | 227 | 30031671 | ||
BC132465.1 | - | 2 | 235 | 40130742 | ||
BC132467.1 | - | 2 | 235 | 40130744 | ||
BC145286.1 | - | 2 | 259 | 9053840 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
