siRNA Details
Gene Symbol: Strada | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252448.1 | NP_001239377.1 | 9, 10 | 943 | ||
NM_001252449.1 | NP_001239378.1 | 7, 8 | 832 | |||
NM_028126.3 | NP_082402.1 | 7, 8 | 832 | |||
GenBank mRNA | AK011451.1 | - | 7, 8 | 778 | ||
AK138358.1 | - | 9, 10 | 944 | |||
AY341872.1 | - | 7, 8 | 791 | |||
AY341878.1 | - | 7, 8 | 791 | |||
BC052063.1 | - | 839 | ||||
BC058517.1 | - | 7, 8 | 733 | 5717255 |
