siRNA Details
Gene Symbol: Tmem163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028135.2 | NP_082411.1 | 7 | 1101 | ||
GenBank mRNA | AK011522.1 | - | 7 | 846 | ||
AK042650.1 | - | 7 | 850 | |||
AK047006.1 | - | 7 | 844 | |||
BC046234.1 | - | 1101 | 6816957 | |||
BC059048.1 | - | 1101 | 6816957 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
