siRNA Details
Gene Symbol: Trim29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023655.2 | NP_076144.2 | 4 | 1263 | ||
GenBank mRNA | AF230390.1 | - | 2 | 136 | ||
AK013219.1 | - | 3 | 314 | |||
AK028448.1 | - | 4 | 496 | |||
BB612224.1 | - | 4 | 493 | |||
BC006699.1 | - | 4 | 994 | |||
BC027353.1 | - | 4 | 1178 | 3988926 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
