siRNA Details
Gene Symbol: Fbxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178624.6 | NP_848739.1 | 6 | 366 | ||
GenBank mRNA | AK013142.1 | - | 7 | 500 | ||
AK039010.1 | - | 6 | 366 | |||
AK044693.1 | - | 6 | 363 | |||
AK045742.1 | - | 6 | 353 | |||
AK089994.1 | - | 6 | 342 | |||
AK141177.1 | - | 6 | 356 | |||
AK163472.1 | - | 6 | 362 | |||
BC051530.1 | - | 2 | 180 | |||
BC096582.1 | - | 6 | 352 | 3963599 | ||
BC145665.1 | - | 6 | 346 | 40130968 | ||
BC145998.1 | - | 6 | 346 | 40131301 |

More Information
Other Species Targeted

Gene Ontology Categories:
