siRNA Details
Gene Symbol: Zfp661 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111029.1 | NP_001104499.1 | 5 | 448 | ||
NM_028141.3 | NP_082417.3 | 6 | 665 | |||
GenBank mRNA | AK033122.1 | - | 5 | 450 | ||
AK134651.1 | - | 6 | 1718 | |||
BC052876.1 | - | 6 | 669 | 30047253 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
