siRNA Details
Gene Symbol: Dbndd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028146.4 | NP_082422.3 | 3 | 476 | ||
GenBank mRNA | AK013173.1 | - | 3 | 375 | ||
AK028187.1 | - | 3 | 413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
