siRNA Details
Gene Symbol: MUC5AC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304359.1 | NP_001291288.1 | 5541 | |||
GenBank mRNA | AJ001402.1 | - | 1 | 427 | ||
AJ298317.1 | - | 31 | 5532 | |||
U06711.1 | - | 1 | 308 | |||
X81649.1 | - | 8 | 2483 | |||
Z34277.1 | - | 1 | 331 | |||
Z34279.1 | - | 1 | 22 | |||
Z48314.1 | - | 1 | 36 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
