siRNA Details
Gene Symbol: Eml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162996.1 | NP_001156468.1 | 16 | 1619 | ||
NM_028153.1 | NP_082429.1 | 16 | 1676 | |||
GenBank mRNA | AK005562.1 | - | 16 | 1642 | ||
BC055476.1 | - | 16 | 1619 | 4218133 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
