siRNA Details
Gene Symbol: 1700001P01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028156.2 | NP_082432.2 | 2 | 320 | ||
GenBank mRNA | AK005619.1 | - | 2 | 284 | ||
BC048607.1 | - | 2 | 320 | 6741651 | ||
CA466212.1 | - | 288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
