siRNA Details
Gene Symbol: Zfp558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028935.1 | NP_083211.1 | 8 | 992 | ||
GenBank mRNA | AK005691.1 | - | 2 | 217 | ||
AK016584.1 | - | 10 | 2646 | |||
BC048749.1 | - | 7 | 700 |
