siRNA Details
Gene Symbol: Psg21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027403.4 | NP_081679.2 | 2 | 406 | ||
GenBank mRNA | AK005492.1 | - | 2 | 404 | ||
AK005540.1 | - | 2 | 401 | |||
AK005545.1 | - | 2 | 404 | |||
AK131670.1 | - | 2 | 404 | |||
BC100324.1 | - | 2 | 383 | 30715195 | ||
BC100325.1 | - | 2 | 383 | 30787148 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
