siRNA Details
Gene Symbol: 1600014C10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085385.1 | NP_001078854.1 | 3 | 574 | ||
NM_028166.3 | NP_082442.1 | 3 | 382 | |||
GenBank mRNA | AK005449.1 | - | 3 | 354 | ||
AK032342.1 | - | 3 | 353 | |||
AK146474.1 | - | 3 | 362 | |||
BC010476.1 | - | 3 | 338 | 3673971 | ||
BC019834.1 | - | 3 | 505 | 5123719 | ||
BC085480.1 | - | 3 | 320 | 6401538 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
