siRNA Details
Gene Symbol: Kcnk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029911.4 | NP_084187.2 | 7 | 1370 | ||
GenBank mRNA | AK031904.1 | - | 7 | 1301 | ||
AK036066.1 | - | 7 | 1644 | |||
AK052327.1 | - | 1 | 233 | |||
AK082153.1 | - | 7 | 1371 | |||
BC132487.1 | - | 7 | 1312 | 40130764 | ||
BC137869.1 | - | 7 | 1312 | 8860888 | ||
DQ185134.1 | - | 7 | 1267 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
