siRNA Details
Gene Symbol: Cda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028176.1 | NP_082452.1 | 3 | 325 | ||
GenBank mRNA | AK008793.1 | - | 3 | 331 | ||
BC022649.1 | - | 1 | 48 | |||
BC050114.1 | - | 3 | 325 | 6433052 | ||
BC056179.1 | - | 2 | 161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
