siRNA Details
Gene Symbol: Smim24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008832.1 | - | 1 | 250 | ||
AK030337.1 | - | 4 | 653 | |||
BC036156.1 | - | 4 | 643 | |||
BC036160.1 | - | 4 | 604 | |||
BC100511.1 | - | 3 | 670 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
