siRNA Details
Gene Symbol: 1810062G17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028183.1 | NP_082459.1 | 2 | 317 | ||
GenBank mRNA | AK007933.1 | - | 2 | 317 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
