siRNA Details
Gene Symbol: Plekhf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024413.2 | NP_077724.2 | 2 | 1511 | ||
GenBank mRNA | AK007489.1 | - | 1 | 919 | ||
AK143976.1 | - | 1500 | ||||
AK157990.1 | - | 2 | 1500 | |||
AK170283.1 | - | 2 | 1510 | |||
BC002120.1 | - | 1 | 1477 | 3491671 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
