siRNA Details
Gene Symbol: 1810041L15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163145.1 | NP_001156617.1 | 3 | 232 | ||
GenBank mRNA | AK129415.1 | - | 4 | 455 | ||
BC062953.1 | - | 3 | 232 | 6856297 | ||
BC119234.1 | - | 3 | 194 | 8733983 | ||
BC120551.1 | - | 3 | 194 | 40129474 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
