siRNA Details
Gene Symbol: Cyp2c65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028191.2 | NP_082467.2 | 4 | 608 | ||
GenBank mRNA | AK008688.1 | - | 4 | 538 | ||
AY227733.1 | - | 4 | 599 | |||
BC051089.1 | - | 4 | 592 | |||
BC117531.1 | - | 4 | 591 | 40106344 | ||
BC118514.1 | - | 4 | 592 | 40106342 |
