siRNA Details
Gene Symbol: Kif1bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028197.2 | NP_082473.2 | 4 | 846 | ||
GenBank mRNA | AK035659.1 | - | 4 | 834 | ||
AK129320.1 | - | 4 | 831 | |||
AK141324.1 | - | 4 | 854 | |||
AK149629.1 | - | 4 | 783 | |||
AK158730.1 | - | 5 | 581 | |||
AK161007.1 | - | 832 | ||||
AK206480.1 | - | 1 | 20 | |||
AK214549.1 | - | 1 | 20 | |||
AK216082.1 | - | 1 | 98 | |||
BC046485.1 | - | 4 | 817 | 5350638 | ||
BC055307.1 | - | 4 | 800 | 6493547 | ||
BC090974.1 | - | 4 | 800 | 6774777 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
